Please login to your online account to display your discounted pricing

anti-Akt1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acid residues 464-477 of human Akt1.
Antigen Akt1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056631

 
novus biologicals canada
NB10056631
Unlabeled 0.2mL Each for $255.80

NB10056631T

 
novus biologicals canada
NB10056631SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acid residues 464-477 of human Akt1.
Antigen Akt1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Test Specificity Cross reacts with Human. Predicted to react with Mouse, Frog and Chicken.
Gene Accession No. NP_001014432
Gene ID (Entrez) 207
Formulation PBS and 0.1mg/mL BSA
Concentration 0.1mg/mL