Please login to your online account to display your discounted pricing

anti-Akt1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056631SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056631T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 207
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Cross Reactivity Human
Formulation PBS and 0.1mg/mL BSA
Concentration 0.1mg/mL
Gene Accession No. NP_001014432
Test Specificity Cross reacts with Human. Predicted to react with Mouse, Frog and Chicken.
Conjugate Unlabeled
Format Purified
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acid residues 464-477 of human Akt1.
Antigen Akt1
Quantity 0.05mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Regulatory Status RUO
Host Species Rabbit