Please login to your online account to display your discounted pricing

anti-AKAP95/AKAP8, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179139

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP179139

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O43823
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 10270
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Antiserum
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Antigen AKAP95/AKAP8
Quantity 0.1mL
Immunogen Human AKAP95/AKAP8 peptide within amino acids 650 to 692 [Swiss-Prot# O43823].