Please login to your online account to display your discounted pricing

anti-AKAP95/AKAP8, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Human AKAP95/AKAP8 peptide within amino acids 650 to 692 [Swiss-Prot# O43823].
Antigen AKAP95/AKAP8
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP179139

 
novus biologicals canada
NBP179139
Unlabeled 0.1mL Each for $309.05

NBP179139SS

 
novus biologicals canada
NBP179139SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Human AKAP95/AKAP8 peptide within amino acids 650 to 692 [Swiss-Prot# O43823].
Antigen AKAP95/AKAP8
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. O43823
Gene ID (Entrez) 10270