Please login to your online account to display your discounted pricing

anti-AIMP2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 140-190 from human AIMP2 was used as the immunogen for this antibody.
Antigen AIMP2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227094

 
novus biologicals canada
NBP227094
Unlabeled 0.1mg Each for $255.80

NBP227094SS

 
novus biologicals canada
NBP227094SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 140-190 from human AIMP2 was used as the immunogen for this antibody.
Antigen AIMP2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, crab-eating macaque, rhesus monkey, white-cheeked gibbon, chimpanzee, and orangutan, and 94% homologous in new world monkey.
Gene Accession No. Q13155
Gene ID (Entrez) 7965
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL