Please login to your online account to display your discounted pricing

anti-AIF-1/Iba1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001028SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001028SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 199
Formulation Tris saline containing (pH 7.3) with 0.5% BSA
Concentration 0.5mg/mL
Gene Accession No. P55008
Test Specificity This antibody is expected to recognize isoform 1 (NP_116573.1) and isoform 3 (NP_001614.3). Expected from sequence similarity: Pig.
Conjugate Unlabeled
Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Peptide ELISA
Applications Western Blotting
Immunogen Synthetic peptide: TGPPAKKAISELP
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Host Species Goat
Monoclonal or Polyclonal Polyclonal
Quantity 0.02mg
Regulatory Status RUO
Antigen AIF-1/Iba1