Please login to your online account to display your discounted pricing

anti-AHR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227296SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227296SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 196
Monoclonal or Polyclonal Polyclonal
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Host Species Rabbit
Concentration 0.5mg/mL
Gene Accession No. P35869
Quantity 0.025mg
Antigen AHR
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Format Purified
Conjugate Unlabeled
Immunogen DLHGISQEFPYkSEMDSMPc (aa677-695).
Primary or Secondary Primary
Cross Reactivity Human