Please login to your online account to display your discounted pricing

anti-AHR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227296

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227296

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene ID (Entrez) 196
Conjugate Unlabeled
Concentration 0.5mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Gene Accession No. P35869
Host Species Rabbit
Quantity 0.1mg
Applications Immunohistochemistry (Paraffin)
Format Purified
Antigen AHR
Immunogen DLHGISQEFPYkSEMDSMPc (aa677-695).
Cross Reactivity Human