Please login to your online account to display your discounted pricing

anti-AHR, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen DLHGISQEFPYkSEMDSMPc (aa677-695).
Antigen AHR
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227296SS

 
novus biologicals canada
NBP227296SS
Unlabeled 0.025mg Each for $109.00

NBP227296

 
novus biologicals canada
NBP227296
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen DLHGISQEFPYkSEMDSMPc (aa677-695).
Antigen AHR
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Gene Accession No. P35869
Gene ID (Entrez) 196
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL