Please login to your online account to display your discounted pricing

anti-AGPAT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002389SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002389SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q86UL3
Conjugate Unlabeled
Antigen AGPAT6
Gene ID (Entrez) 137964
Test Specificity Human, monkey and mouse. Predicted to react with bovine, porcine, and rat based on 100% sequence homology.
Formulation Tris-glycine, 150mM NaCl
Concentration 0.81mg/mL
Regulatory Status RUO
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Quantity 0.025mL
Format Affinity Purified
Immunogen A synthetic peptide made to an N-terminal portion of human AGPAT6 (between residues 1-100). [Swiss-Prot Q86UL3]
Primary or Secondary Primary
Host Species Rabbit