Please login to your online account to display your discounted pricing

anti-AGPAT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002389

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002389

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 0.81mg/mL
Quantity 0.1mL
Gene ID (Entrez) 137964
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Gene Accession No. Q86UL3
Applications Western Blotting
Test Specificity Human, monkey and mouse. Predicted to react with bovine, porcine, and rat based on 100% sequence homology.
Regulatory Status RUO
Conjugate Unlabeled
Format Affinity Purified
Immunogen A synthetic peptide made to an N-terminal portion of human AGPAT6 (between residues 1-100). [Swiss-Prot Q86UL3]
Antigen AGPAT6
Host Species Rabbit