Please login to your online account to display your discounted pricing

anti-AGPAT6, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of human AGPAT6 (between residues 400-456). [Swiss-Prot Q86UL3]
Antigen AGPAT6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002390

 
novus biologicals canada
NB1002390
Unlabeled 0.1mL Each for $309.05

NB1002390SS

 
novus biologicals canada
NB1002390SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of human AGPAT6 (between residues 400-456). [Swiss-Prot Q86UL3]
Antigen AGPAT6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human, monkey and mouse. Predicted to react with rat, bovine, and porcine based on 100% sequence homology.
Gene Accession No. Q86UL3
Gene ID (Entrez) 137964
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL