Please login to your online account to display your discounted pricing

anti-AGPAT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002390SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002390SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene Accession No. Q86UL3
Gene ID (Entrez) 137964
Conjugate Unlabeled
Quantity 0.025mL
Concentration 1mg/mL
Test Specificity Human, monkey and mouse. Predicted to react with rat, bovine, and porcine based on 100% sequence homology.
Formulation Tris-glycine, 150mM NaCl
Format Affinity Purified
Antigen AGPAT6
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Regulatory Status RUO
Immunogen A synthetic peptide made to a C-terminal portion of human AGPAT6 (between residues 400-456). [Swiss-Prot Q86UL3]
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal