Please login to your online account to display your discounted pricing

anti-AGPAT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002390

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002390

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Test Specificity Human, monkey and mouse. Predicted to react with rat, bovine, and porcine based on 100% sequence homology.
Regulatory Status RUO
Applications Western Blotting
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Gene ID (Entrez) 137964
Gene Accession No. Q86UL3
Antigen AGPAT6
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Immunogen A synthetic peptide made to a C-terminal portion of human AGPAT6 (between residues 400-456). [Swiss-Prot Q86UL3]
Format Affinity Purified
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary