Please login to your online account to display your discounted pricing

anti-Ago2/eIF2C2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056198

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056198

  • / Each
 

Description & Specifications

Specifications

Antigen Ago2/eIF2C2
Applications Electron Microscopy
Applications Peptide ELISA
Conjugate Unlabeled
Cross Reactivity Drosophila
Cross Reactivity Human
Format Antisera
Formulation 50% glycerol
Host Species Rabbit
Immunogen Amino acids 361-378 (TSTMIRATARSAPDRQEE) of human Argonaut 2 protein were used as the immunogen. Genbank accession number: NP_036286.
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 27161
Test Specificity The amino acid sequence used is 100% homologous in human Argonaut 2 (Gene ID: 27161, UniProt: Q9UKV8), Argonaut 3 (Gene ID: 192669, UniProt: Q9H9G7), and Argonaut 4 (Gene ID: 192670, UniProt: Q9HCK5) and 94% homologous in Argonaut 1. The immunizing sequence is highly conserved between various isoforms (isoforms 1, 2, 3 and 4, some having yet additional isoforms) and species mouse, rat, cow, dog, xenopus, rabbit, chimp and chicken. Human and Drosophila. Predicted cross-reactivity based on sequence identity: Mouse, Rat, Cow, Chicken.
Target Species Drosophila
Target Species Human