Please login to your online account to display your discounted pricing

anti-Ago2/eIF2C2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 361-378 (TSTMIRATARSAPDRQEE) of human Argonaut 2 protein were used as the immunogen. Genbank accession number: NP_036286.
Antigen Ago2/eIF2C2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056198T

 
novus biologicals canada
NB10056198SS
Unlabeled 0.025mL Each for $139.00

NB10056198

 
novus biologicals canada
NB10056198
Unlabeled 0.1mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 361-378 (TSTMIRATARSAPDRQEE) of human Argonaut 2 protein were used as the immunogen. Genbank accession number: NP_036286.
Antigen Ago2/eIF2C2
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Electron Microscopy
Applications Peptide ELISA
Test Specificity The amino acid sequence used is 100% homologous in human Argonaut 2 (Gene ID: 27161, UniProt: Q9UKV8), Argonaut 3 (Gene ID: 192669, UniProt: Q9H9G7), and Argonaut 4 (Gene ID: 192670, UniProt: Q9HCK5) and 94% homologous in Argonaut 1. The immunizing sequence is highly conserved between various isoforms (isoforms 1, 2, 3 and 4, some having yet additional isoforms) and species mouse, rat, cow, dog, xenopus, rabbit, chimp and chicken. Human and Drosophila. Predicted cross-reactivity based on sequence identity: Mouse, Rat, Cow, Chicken.
Gene ID (Entrez) 27161
Formulation 50% glycerol