Please login to your online account to display your discounted pricing

anti-AG-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11017780

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11017780

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. O95994
Test Specificity Human. Immunogen sequence has 94% homology to bovine and 88% homology to mouse.
Host Species Rabbit
Formulation PBS, 30% glycerol
Gene ID (Entrez) 10551
Conjugate Unlabeled
Immunogen A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt No. O95994]
Concentration 1.0mg/mL
Antigen AG-2
Quantity 0.1mL
Cross Reactivity Human
Primary or Secondary Primary
Format Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO