Please login to your online account to display your discounted pricing

anti-AG-2, FITC, Polyclonal, Novus Biologicals™

$406.00 - $406.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt No. O95994]
Antigen AG-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP105936F

 
novus biologicals canada
NBP105936F
FITC 0.1mL Each for $406.00

NB11017780F

 
novus biologicals canada
NB11017780F
FITC 0.05mL Each for $406.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt No. O95994]
Antigen AG-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Human. Immunogen sequence has 94% homology to bovine and 88% homology to mouse.
Gene Accession No. O95994
Gene ID (Entrez) 10551
Formulation 50mM sodium borate