Please login to your online account to display your discounted pricing

anti-AG-2, FITC, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11017780F

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11017780F

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate FITC
Gene Accession No. O95994
Formulation 50mM sodium borate
Quantity 0.05mL
Gene ID (Entrez) 10551
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Human. Immunogen sequence has 94% homology to bovine and 88% homology to mouse.
Antigen AG-2
Regulatory Status RUO
Host Species Rabbit
Format Affinity Purified
Immunogen A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt No. O95994]
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal