missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-AG-2, DyLight 650, Polyclonal, Novus Biologicals™

$406.00 - $406.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt No. O95994]
Antigen AG-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP105936C Novus Biologicals Canada
NBP105936C
0.1mL Each for $406.00
NB11017780C Novus Biologicals Canada
NB11017780C
0.05mL Each for $406.00
Specifications Description & Specifications

Specifications

Rabbit
AG-2
Affinity Purified
RUO
Human. Immunogen sequence has 94% homology to bovine and 88% homology to mouse.
10551
A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt No. O95994]
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin)
O95994
50mM sodium borate