Please login to your online account to display your discounted pricing

anti-AG-2, DyLight 650, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP105936C

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP105936C

  • / Each
 

Description & Specifications

Specifications

Antigen AG-2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Conjugate DyLight 650
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation 50mM sodium borate
Gene Accession No. O95994
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of mouse AGR2 (within residues 1-50). [Swiss-Prot No. O88312]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10551
Target Species Bovine
Target Species Human
Target Species Murine