Please login to your online account to display your discounted pricing

anti-AG-2, DyLight 550, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11017780R

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11017780R

  • / Each
 

Description & Specifications

Specifications

Formulation 50mM sodium borate
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10551
Test Specificity Human. Immunogen sequence has 94% homology to bovine and 88% homology to mouse.
Antigen AG-2
Gene Accession No. O95994
Immunogen A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt No. O95994]
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Cross Reactivity Human
Conjugate DyLight 550
Format Affinity Purified
Quantity 0.05mL