Please login to your online account to display your discounted pricing

anti-AG-2, DyLight 488, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11017780G

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11017780G

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10551
Conjugate DyLight 488
Formulation 50mM sodium borate
Primary or Secondary Primary
Gene Accession No. O95994
Host Species Rabbit
Test Specificity Human. Immunogen sequence has 94% homology to bovine and 88% homology to mouse.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Regulatory Status RUO
Format Affinity Purified
Antigen AG-2
Quantity 0.05mL
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt No. O95994]