Please login to your online account to display your discounted pricing

anti-AG-2, DyLight 488, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP105936G

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP105936G

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Gene Accession No. O95994
Conjugate DyLight 488
Formulation 50mM sodium borate
Gene ID (Entrez) 10551
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Regulatory Status RUO
Immunogen Synthetic peptide made to an N-terminal portion of mouse AGR2 (within residues 1-50). [Swiss-Prot No. O88312]
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Quantity 0.1mL
Antigen AG-2