Please login to your online account to display your discounted pricing

anti-AG-2, Biotin, Polyclonal, Novus Biologicals™

$406.00 - $406.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of mouse AGR2 (within residues 1-50). [Swiss-Prot No. O88312]
Antigen AG-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP105936B

 
novus biologicals canada
NBP105936B
Biotin 0.1mL Each for $406.00

NB11017780B

 
novus biologicals canada
NB11017780B
Biotin 0.05mL Each for $406.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of mouse AGR2 (within residues 1-50). [Swiss-Prot No. O88312]
Antigen AG-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene Accession No. O95994
Gene ID (Entrez) 10551
Formulation PBS