missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adiponectin Receptor 2 Polyclonal Antibody, Invitrogen™

Adiponectin Receptor 2 Antibody, PA1-1071, from Invitrogen™. Species Reactivity: Mouse, Rat; Applications: Western Blot.

Manufacturer:  Pierce Biotechnology Inc PA11071

Catalog No. PA11071


 

Specifications

Description

Adiponectin Receptor 2 Antibody, PA1-1071, from Invitrogen. Species Reactivity: Mouse, Rat; Applications: Western Blot.

Adiponectin (also known as 30-kDa adipocyte complement-related protein; Acrp30) is a hormone secreted by adipocytes that acts as an antidiabetic and anti-atherogenic adipokine. Levels of adiponectin in the blood are decreased under conditions of obesity, insulin resistance and type 2 diabetes. Administration of adiponectin causes glucose-lowering effects and ameliorates insulin resistance in mice. Conversely, adiponectin-deficient mice exhibit insulin resistance and diabetes. This insulin-sensitizing effect of adiponectin seems to be mediated by an increase in fatty-acid oxidation through activation of AMP kinase and PPAR- . Here we report the cloning of complementary DNAs encoding adiponectin receptors 1 and 2 (AdipoR1 and AdipoR2) by expression cloning. AdipoR1 is abundantly expressed in skeletal muscle, whereas AdipoR2 is predominantly expressed in the liver. These two adiponectin receptors are predicted to contain seven transmembrane domains, but are structurally and functionally distinct from G-protein-coupled receptors. Expression of AdipoR1/R2 or suppression of AdipoR1/R2 expression by small-interfering RNA supports our conclusion that they serve as receptors for globular and full-length adiponectin, and that they mediate increased AMP kinase and PPAR- ligand activities, as well as fatty-acid oxidation and glucose uptake by adiponectin.
Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues D M F A T P Q Y R G V R of mouse Adiponectin Receptor 2.
Formulation: 100μg purified antibody (1mg/mL) in PBS with 1mg/mL BSA and 0.05% sodium azide
Synonyms: ADIPOR2, ACDCR2, Adiponectin receptor protein 2, FLJ21432, MGC4640, PAQR2, Progestin and adipoQ receptor family member II
Uniprot Accession: Q8BQS5
Entrez GeneID: 68465
Gene Symbol: ADIPOR2
PA1-1071 detects Adiponectin Receptor 2 from mouse and rat samples. PA1-1071 has been successfully used in Western blot applications. By Western blot, this antibody detects a single ∽43 kDa band representing Adiponectin Receptor 2 from mouse liver. The PA1-1071 immunogen is a synthetic peptide corresponding to residues D M F A T P Q Y R G V R of mouse Adiponectin Receptor 2.
References:
Test: WB, Species: Ms, Reference: A peptide-modified chitosan-collagen hydrogel for cardiac cell culture and delivery. (2012 Mar) Acta Biomater, Vol: 8, 3, 1022-36, Reis LA, Chiu LL, Liang Y, Hyunh K, Momen A, Radisic M; Pubmed ID: 22155066
Description & Specifications

Specifications

Adiponectin Receptor 2
Unlabeled
Purified
Q8BQS5
ADIPOR2
Synthetic peptide corresponding to residues D M F A T P Q Y R G V R of mouse Adiponectin Receptor 2.
100μg
Avoid Freeze/Thaw Cycles
Polyclonal
Murine,Rat
Western Blot
Mouse,Rat
100μg purified antibody (1mg/mL) in PBS with 1mg/mL BSA and 0.05% sodium azide
ADIPOR2, ACDCR2, Adiponectin receptor protein 2, FLJ21432, MGC4640, PAQR2, Progestin and adipoQ receptor family member II
Rabbit
IgG
Mouse, Rat
Primary
68465
Promotions

Promotions

  • Purchase any three Invitrogen™ Antibodies and receive a fourth antibody at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.

    06/30/18