Please login to your online account to display your discounted pricing

anti-Adiponectin/Acrp30, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10065810SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10065810T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity This recognises an epitope within the C-terminal region (CT) of the 244 amino acid protein Adiponectin.
Gene ID (Entrez) 9370
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. Q15848
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal sequence of human Adiponectin (between amino acids 200-244) [UniProt Q15848]
Quantity 0.025mg
Regulatory Status RUO
Format Affinity Purified
Antigen Adiponectin/Acrp30