Please login to your online account to display your discounted pricing

anti-Adiponectin/Acrp30, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10065810

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10065810

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q15848
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Concentration 1.0mg/mL
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity This recognises an epitope within the C-terminal region (CT) of the 244 amino acid protein Adiponectin.
Conjugate Unlabeled
Formulation PBS
Gene ID (Entrez) 9370
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen A synthetic peptide made to a C-terminal sequence of human Adiponectin (between amino acids 200-244) [UniProt Q15848]
Host Species Rabbit
Regulatory Status RUO
Quantity 0.1mg
Antigen Adiponectin/Acrp30