Please login to your online account to display your discounted pricing

anti-Adiponectin/Acrp30, Polyclonal, Novus Biologicals™

$139.00 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal sequence of human Adiponectin (between amino acids 200-244) [UniProt Q15848]
Antigen Adiponectin/Acrp30
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10065810

 
novus biologicals canada
NB10065810
Unlabeled 0.1mg Each for $384.00

NB10065810T

 
novus biologicals canada
NB10065810SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal sequence of human Adiponectin (between amino acids 200-244) [UniProt Q15848]
Antigen Adiponectin/Acrp30
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This recognises an epitope within the C-terminal region (CT) of the 244 amino acid protein Adiponectin.
Gene Accession No. Q15848
Gene ID (Entrez) 9370
Formulation PBS
Concentration 1.0mg/mL