missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ADFP Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA116971
Description
This antibody is predicted to react with rat based on 84% sequence homology.
Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.Specifications
ADFP | |
Polyclonal | |
Unconjugated | |
PLIN2 | |
Adipophilin antibody, adipose differentiation-related protein antibody, ADRP antibody | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11520, 123 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin),Western Blot | |
1.23 mg/mL | |
PBS with 30% glycerol and 0.05% sodium azide | |
Q99541, P43883 | |
PLIN2 | |
Synthetic peptide corresponding to the internal region (within residues 150-250) of human ADFP. | |
100 μL | |
Primary | |
Human,Mouse | |
Antibody | |
IgG |