Please login to your online account to display your discounted pricing

anti-Activin RIA/ALK-2/Activin Receptor Type 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056432

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056432

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin.
Gene Accession No. NP_001104537.1
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 90
Regulatory Status RUO
Test Specificity In human kidney, additional bands are seen at approximately 27 and 14 kD.
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Applications Western Blotting
Conjugate Unlabeled
Format Purified
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 475-525 of human ACVR1 was used as immunogen.
Quantity 0.1mg
Antigen Activin RIA/ALK-2/Activin Receptor Type 1