Please login to your online account to display your discounted pricing

anti-Activin RIA/ALK-2/Activin Receptor Type 1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 475-525 of human ACVR1 was used as immunogen.
Antigen Activin RIA/ALK-2/Activin Receptor Type 1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056432

 
novus biologicals canada
NB10056432
Unlabeled 0.1mg Each for $255.80

NB10056432T

 
novus biologicals canada
NB10056432SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 475-525 of human ACVR1 was used as immunogen.
Antigen Activin RIA/ALK-2/Activin Receptor Type 1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity In human kidney, additional bands are seen at approximately 27 and 14 kD.
Gene Accession No. NP_001104537.1
Gene ID (Entrez) 90
Formulation PBS containing 0.2% gelatin.
Concentration 0.5mg/mL