Please login to your online account to display your discounted pricing

anti-Activin RIA/ALK-2/Activin Receptor Type 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056432SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056432T

  • / Each
 

Description & Specifications

Specifications

Antigen Activin RIA/ALK-2/Activin Receptor Type 1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.2% gelatin.
Gene Accession No. NP_001104537.1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 475-525 of human ACVR1 was used as immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 90
Test Specificity In human kidney, additional bands are seen at approximately 27 and 14 kD.
Target Species Human
Target Species Murine
Target Species Rat