Please login to your online account to display your discounted pricing

anti-Actin Gamma 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600533SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600533SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P63261
Monoclonal or Polyclonal Polyclonal
Concentration 0.2mg/mL
Format Affinity Purified
Formulation TBS with 0.1% BSA
Gene ID (Entrez) 71
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Immunogen The epitope recognized by this antibody maps to the N-terminus of human Actin Gamma 1 [UniProt No. P63261]. The N-terminus of Actin Gamma 1 is highly conserved with Beta Actin and preliminary indications are that NB600-533 also recognizes Beta Actin (GeneID 60).
Host Species Rabbit
Antigen Actin Gamma 1
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Conjugate Unlabeled