Please login to your online account to display your discounted pricing

anti-Actin Gamma 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600533

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600533

  • / Each
 

Description & Specifications

Specifications

Antigen Actin Gamma 1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation TBS with 0.1% BSA
Gene Accession No. P63261
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to the N-terminus of human Actin Gamma 1 [UniProt No. P63261]. The N-terminus of Actin Gamma 1 is highly conserved with Beta Actin and preliminary indications are that NB600-533 also recognizes Beta Actin (GeneID 60).
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 71
Target Species Human
Target Species Murine