Please login to your online account to display your discounted pricing

anti-Actin Gamma 1, Polyclonal, Novus Biologicals™

$160.00 - $362.00

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to the N-terminus of human Actin Gamma 1 [UniProt No. P63261]. The N-terminus of Actin Gamma 1 is highly conserved with Beta Actin and preliminary indications are that NB600-533 also recognizes Beta Actin (GeneID 60).
Antigen Actin Gamma 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600533

 
novus biologicals canada
NB600533
Unlabeled 0.1mL Each for $362.00

NB600533SS

 
novus biologicals canada
NB600533SS
Unlabeled 0.025mL Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to the N-terminus of human Actin Gamma 1 [UniProt No. P63261]. The N-terminus of Actin Gamma 1 is highly conserved with Beta Actin and preliminary indications are that NB600-533 also recognizes Beta Actin (GeneID 60).
Antigen Actin Gamma 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Gene Accession No. P63261
Gene ID (Entrez) 71
Formulation TBS with 0.1% BSA
Concentration 0.2mg/mL