Please login to your online account to display your discounted pricing

anti-Actin Alpha 2 Smooth Muscle, Polyclonal, Novus Biologicals™

$110.17 - $409.37

Specifications

Host Species Rabbit
Immunogen Synthetic peptide corresponding to N-terminus of human alpha smooth muscle isoform of actin.
Antigen Actin Alpha 2 Smooth Muscle
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600531

 
novus biologicals canada
NB600531
Unlabeled 0.5mL Each for $329.00

NB600531SS

 
novus biologicals canada
NB600531SS
Unlabeled 0.02mL Each for $110.17

NB6005311ML

 
novus biologicals canada
NB6005311ML
Unlabeled 1mL Each for $409.37
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide corresponding to N-terminus of human alpha smooth muscle isoform of actin.
Antigen Actin Alpha 2 Smooth Muscle
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells.
Gene Accession No. P62736
Gene ID (Entrez) 59
Formulation PBS/1% BSA buffer pH 7.6
Concentration 1.0mg/mL