Please login to your online account to display your discounted pricing

anti-Actin Alpha 2 Smooth Muscle, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6005311ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6005311ML

  • / Each
 

Description & Specifications

Specifications

Formulation PBS/1% BSA buffer pH 7.6
Test Specificity This stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells.
Gene Accession No. P62736
Monoclonal or Polyclonal Polyclonal
Concentration 1.0mg/mL
Gene ID (Entrez) 59
Conjugate Unlabeled
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Host Species Rabbit
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 1mL
Antigen Actin Alpha 2 Smooth Muscle
Primary or Secondary Primary
Immunogen Synthetic peptide corresponding to N-terminus of human alpha smooth muscle isoform of actin.