Please login to your online account to display your discounted pricing

anti-Actin Alpha 2 Smooth Muscle, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600531SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600531SS

  • / Each
 

Description & Specifications

Specifications

Antigen Actin Alpha 2 Smooth Muscle
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS/1% BSA buffer pH 7.6
Gene Accession No. P62736
Host Species Rabbit
Immunogen Synthetic peptide corresponding to N-terminus of human alpha smooth muscle isoform of actin.
Quantity 0.02mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 59
Test Specificity This stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells.
Target Species Human
Target Species Murine
Target Species Rat