Please login to your online account to display your discounted pricing

anti-Actin Alpha 2 Smooth Muscle, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600531

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600531

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 59
Conjugate Unlabeled
Formulation PBS/1% BSA buffer pH 7.6
Test Specificity This stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells.
Gene Accession No. P62736
Concentration 1.0mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen Actin Alpha 2 Smooth Muscle
Immunogen Synthetic peptide corresponding to N-terminus of human alpha smooth muscle isoform of actin.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Quantity 0.5mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit