Acetyl Histone H4 (Lys12) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer: Emd Millipore Corporation 061352I
Description
This Anti-acetyl Histone H4 (Lys12) Antibody is validated for use in western blotting, immunocytochemistry and dot blot for the detection of acetyl Histone H4 (Lys12).Specifications
acetyl Histone H4 (Lys12) | |
0.5mg/mL | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HIST1H4A; H4/A; H4FA; HIST1H4B; H4/I; H4FI; HIST1H4C; H4/G; H4FG; HIST1H4D; H4/B; H4FB; HIST1H4E; H4/J; H4FJ; HIST1H4F; H4/C; H4FC; HIST1H4H; H4/H; H4FH; HIST1H4I; H4/M; H4FM; HIST1H4J; H4/E; H4FE; HIST1H4K; H4/D; H4FD; HIST1H4L; H4/K; H4FK; HIST2H4A; H4/N; H4F2; H4FN; HIST2H4; HIST2H4B; H4/O; | |
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 acetylated at Lys12. | |
100μg | |
Primary | |
NP_003529 |
Dot Blot,Immunocytochemistry,Western Blot | |
Unconjugated | |
Affinity Purified | |
Rabbit | |
Affinity purified | |
2-8°C for 12 months | |
Polyclonal | |
Human,Murine,Rat |
For Research Use Only