Please login to your online account to display your discounted pricing

anti-acetyl Histone H4 (Lys12), Polyclonal, EMD Millipore™

$113.00 - $454.00

Specifications

Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 acetylated at Lys12.
Antigen acetyl Histone H4 (Lys12)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

06-135-2IMI

 
emd millipore corporation
061352I
Unlabeled 100μg Each for $454.00

06-135-2ISMI

 
emd millipore corporation
061352IS
Unlabeled 10μg Each for $113.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 acetylated at Lys12.
Antigen acetyl Histone H4 (Lys12)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Prrimary
Applications Dot Blot
Applications Immunocytochemistry
Applications Western Blotting
Gene ID (Entrez) NP_003529
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Concentration 0.5mg/mL