Acetyl Histone H4 (Lys12) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  Emd Millipore Corporation 061352IS
 View more versions of this product
Catalog No. 06-135-2ISMI
Description
Description
This Anti-acetyl Histone H4 (Lys12) Antibody is validated for use in western blotting, immunocytochemistry and dot blot for the detection of acetyl Histone H4 (Lys12).
Specifications
Specifications
acetyl Histone H4 (Lys12)
0.5mg/mL
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
HIST1H4A; H4/A; H4FA; HIST1H4B; H4/I; H4FI; HIST1H4C; H4/G; H4FG; HIST1H4D; H4/B; H4FB; HIST1H4E; H4/J; H4FJ; HIST1H4F; H4/C; H4FC; HIST1H4H; H4/H; H4FH; HIST1H4I; H4/M; H4FM; HIST1H4J; H4/E; H4FE; HIST1H4K; H4/D; H4FD; HIST1H4L; H4/K; H4FK; HIST2H4A; H4/N; H4F2; H4FN; HIST2H4; HIST2H4B; H4/O;
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 acetylated at Lys12.
10μg
Primary
NP_003529
Dot Blot,Immunocytochemistry,Western Blot
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human,Murine,Rat