Acetyl-Histone H3 (Lys9) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma 06942S
 View more versions of this product
Catalog No. 06-942-SMI
Description
Description
Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 acetylated on lysine 9. This purified Ab, also known as Anti-H3K9Ac, is specificity verified by dot blot (DB), published in peer reviewed journals, and validated in western blot, Chimmunoprecipitation, ChIP-seq, flow cytometry.
Specifications
Specifications
acetyl-Histone H3 (Lys9)
Polyclonal
Purified rabbit IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%.
Rabbit
Protein A purfied
Primary
Eukaryote, Human, Mouse, Rat
Purified
ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Flow Cytometry, Western Blot
Unconjugated
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
KLH-conjugated, synthetic peptide (ARTKQTA RAcKSTG-C) corresponding to amino acids 1-12 of Histone H3.
20 μg
NM_003493
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
IgG