missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl-Histone H3 (Lys9) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer: Emd Millipore Corporation 06942
Description
Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 acetylated on lysine 9. This purified Ab, also known as Anti-H3K9Ac, is specificity verified by dot blot (DB), published in peer reviewed journals, and validated in western blot, Chimmunoprecipitation, ChIP-seq, flow cytometry.Specifications
acetyl-Histone H3 (Lys9) | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
200μg | |
Primary | |
NP_003520 |
ChIP-sequencing,Chromatin Immunoprecipitation,Dot Blot,Flow Cytometry,Western Blot | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide with 30% glycerol. | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886 | |
KLH-conjugated synthetic peptide (ARTKQTARK(Ac)STG-C) corresponding to amino acids 1-12 of Histone H3 with acetylated Lys9. | |
Protein A purfied | |
−20°C in undiluted aliquotes for up to 12 months | |
Polyclonal | |
Human,Murine,Rat |
For Research Use Only