Please login to your online account to display your discounted pricing

anti-acetyl-Histone H3 (Lys56), Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  07677I

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 07-677-IMI

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Antigen acetyl-Histone H3 (Lys56)
Format Affinity Purified
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys56.
Purification Method Affinity purified
Quantity 200μL
Primary or Secondary Prrimary
Gene ID (Entrez) NP 003520
Cross Reactivity Human
Storage Requirements 2-8°C for 12 months
Conjugate Unlabeled