Please login to your online account to display your discounted pricing

anti-acetyl-Histone H3 (Lys56), Polyclonal, EMD Millipore™

$110.34 - $110.34

Specifications

Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys56.
Antigen acetyl-Histone H3 (Lys56)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

07-677-ISMI

 
emd millipore corporation
07677IS
Unlabeled 20μL Each for $110.34
Description & Specifications

Specifications

Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys56.
Antigen acetyl-Histone H3 (Lys56)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Prrimary
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Western Blotting
Gene ID (Entrez) NP 003520
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.