Please login to your online account to display your discounted pricing

anti-acetyl-Histone H3 (Lys56), Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  07677IS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 07-677-ISMI

  • / Each
 

Description & Specifications

Specifications

Purification Method Affinity purified
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys56.
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Western Blotting
Cross Reactivity Human
Conjugate Unlabeled
Gene ID (Entrez) NP 003520
Host Species Rabbit
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Prrimary
Quantity 20μL
Storage Requirements 2-8°C for 12 months
Format Affinity Purified
Antigen acetyl-Histone H3 (Lys56)
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.