Please login to your online account to display your discounted pricing

anti-acetyl Histone H3 (Lys4), Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  ABE223

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. ABE223MI

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Prrimary
Conjugate Unlabeled
Purification Method Affinity purified
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys4.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Dot Blot
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) NP_003520
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
Quantity 100μL
Storage Requirements 2-8°C for 12 months
Antigen acetyl Histone H3 (Lys4)
Format Affinity Purified
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit