missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl Histone H3 (Lys4) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$118.34 - $474.33

Specifications

Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys4.
Antigen acetyl Histone H3 (Lys4)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ABE223SMI View Documents MilliporeSigma
ABE223S
10μL Each for $118.34
 
ABE223MI View Documents MilliporeSigma
ABE223
100μL Each for $474.33
 
Description

Description

Use Anti-acetyl Histone H3 (Lys4) Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunoprecipitation, DB to detect acetyl Histone H3 (Lys4) also known as Histone H3.1.
Specifications

Specifications

Rabbit
acetyl Histone H3 (Lys4)
Affinity Purified
Dot Blot, Immunoprecipitation, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
KLH-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys4.
Polyclonal
Primary
NP_003520

For Research Use Only